Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxw8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172721.2 | NP_766309.2 | 10 | 1637 | ||
GenBank mRNA | AK031815.1 | - | 10 | 1631 | ||
AK034447.1 | - | 10 | 1565 | |||
AK053794.1 | - | 10 | 1638 | |||
AK134291.1 | - | 10 | 1644 | |||
AK163976.1 | - | 2 | 186 | |||
BC009095.1 | - | 10 | 1590 | 3158218 | ||
BC024091.1 | - | 4 | 513 |