Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naa25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172722.3 | NP_766310.2 | 5, 6 | 502 | ||
GenBank mRNA | AK049325.1 | - | 5, 6 | 503 | ||
AK052060.1 | - | 5, 6 | 490 | |||
AK149758.1 | - | 5, 6 | 503 | |||
AK165175.1 | - | 5, 6 | 493 | |||
AK167405.1 | - | 5, 6 | 520 | |||
AK167491.1 | - | 5, 6 | 520 | |||
BC028868.1 | - | 3, 4 | 282 | |||
BC037595.1 | - | 5, 6 | 483 | |||
BC055480.1 | - | 4, 5 | 404 | |||
BC082314.1 | - | 5, 6 | 488 | |||
BC106182.1 | - | 5, 6 | 477 | |||
BC132172.1 | - | 5, 6 | 500 | 40130449 | ||
BC138093.1 | - | 5, 6 | 500 | 8861112 |