Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naa25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172722.3 | NP_766310.2 | 20 | 2416 | ||
GenBank mRNA | AK041906.1 | - | 6 | 733 | ||
AK049325.1 | - | 20 | 2417 | |||
AK051815.1 | - | 3 | 172 | |||
AK149758.1 | - | 20 | 2417 | |||
AK165175.1 | - | 20 | 2408 | |||
AK167405.1 | - | 20 | 2434 | |||
AK167491.1 | - | 20 | 2434 | |||
BC028868.1 | - | 18 | 2196 | |||
BC037595.1 | - | 20 | 2397 | |||
BC055480.1 | - | 19 | 2318 | |||
BC082314.1 | - | 20 | 2402 | |||
BC106182.1 | - | 20 | 2391 | |||
BC132172.1 | - | 20 | 2414 | 40130449 | ||
BC138093.1 | - | 20 | 2414 | 8861112 |