Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rimbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081388.1 | NP_001074857.1 | 20 | 3639 | ||
GenBank mRNA | AK038814.1 | - | 19 | 3107 | ||
AK044685.1 | - | 19 | 3739 | |||
AK122245.2 | - | 11 | 2452 | |||
AK147417.1 | - | 16 | 3045 | |||
BC151100.1 | - | 16 | 3230 | 9088013 | ||
BC151101.1 | - | 16 | 3230 | 9088014 |