Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mepce | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144913.3 | NP_659162.3 | 1 | 1141 | ||
GenBank mRNA | AK154415.1 | - | 1 | 1104 | ||
BC026876.1 | - | 1 | 613 | 3662118 |