Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pilra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153510.3 | NP_705730.1 | 3 | 801 | ||
GenBank mRNA | AJ400844.1 | - | 699 | |||
AK041413.1 | - | 2 | 1092 | |||
AK041670.1 | - | 3 | 713 | |||
AK170000.1 | - | 3 | 713 | |||
BC115565.2 | - | 686 | 40102015 |