Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276287.1 | NP_001263216.1 | 9 | 1361 | ||
NM_181066.3 | NP_851411.1 | 9 | 1263 | |||
GenBank mRNA | AK043212.1 | - | 9 | 1250 | ||
AK049321.1 | - | 8 | 1160 | |||
AK084987.1 | - | 9 | 1252 | |||
AK085091.1 | - | 9 | 1252 | |||
AK142387.1 | - | 6 | 2519 | |||
AK154448.1 | - | 9 | 1234 | |||
AK170269.1 | - | 9 | 1257 | |||
BC068301.1 | - | 9 | 1327 | 30061848 |