Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173405.2 | NP_775581.1 | 4 | 811 | ||
GenBank mRNA | AJ879912.1 | - | 2 | 452 | ||
AK078332.1 | - | 4 | 811 | |||
AK170176.1 | - | 3 | 785 | |||
BC129841.1 | - | 2 | 594 | 40090727 | ||
BC129842.1 | - | 2 | 546 | 40090729 |