Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap5z1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172725.2 | NP_766313.2 | 16 | 2123 | ||
GenBank mRNA | AK040924.1 | - | 14 | 1675 | ||
AK082759.1 | - | 8 | 950 | |||
AK129135.1 | - | 12 | 3391 | |||
AK146431.1 | - | 16 | 2001 | |||
AK152402.1 | - | 16 | 2001 | |||
AK153787.1 | - | 14 | 3712 | |||
AK185767.1 | - | 3 | 185 | |||
AK187973.1 | - | 3 | 186 | |||
AK215647.1 | - | 3 | 185 | |||
BC057548.1 | - | 16 | 1930 | |||
BC144755.1 | - | 14 | 1606 |