Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap5z1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172725.2 | NP_766313.2 | 2 | 243 | ||
GenBank mRNA | AK129135.1 | - | 2 | 114 | ||
AK146431.1 | - | 2 | 121 | |||
AK152402.1 | - | 2 | 121 | |||
AK153787.1 | - | 2 | 116 | |||
BC057548.1 | - | 2 | 98 | |||
BC082786.1 | - | 2 | 216 | |||
BC099419.1 | - | 2 | 175 | |||
BY125380.1 | - | 2 | 244 |