Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLXNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025179.3 | NP_079455.3 | 18 | 4241 | ||
XM_005273164.2 | XP_005273221.1 | 18 | 4305 | |||
XM_005273165.3 | XP_005273222.1 | 18 | 4305 | |||
GenBank mRNA | AB007932.1 | - | 18 | 3692 | ||
BC009343.2 | - | 1 | 231 | |||
BC132676.1 | - | 18 | 3596 | 40146698 | ||
BC136530.1 | - | 18 | 3596 | 9020520 | ||
X87831.2 | - | 16 | 2146 |