Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289589.1 | NP_001276518.1 | 5 | 723 | ||
NM_001289590.1 | NP_001276519.1 | 5 | 793 | |||
NM_177681.4 | NP_808349.1 | 4 | 627 | |||
GenBank mRNA | AK049650.1 | - | 4 | 583 | ||
BB612291.1 | - | 4 | 627 | |||
BC053080.1 | - | 5 | 632 | 5702966 | ||
BC059836.1 | - | 5 | 704 | 5697949 |