Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033313.3 | NP_001028485.1 | 2 | 181 | ||
GenBank mRNA | AK147170.1 | - | 2 | 182 | ||
AK169233.1 | - | 2 | 114 | |||
BC007162.1 | - | 1 | 81 | |||
BC055927.1 | - | 2 | 95 | |||
BC106168.1 | - | 1 | 69 | |||
BC139204.1 | - | 2 | 134 | 8862226 | ||
BC139205.1 | - | 2 | 134 | 8862227 |