Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnpda1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011937.2 | NP_036067.2 | 6, 7 | 871 | ||
GenBank mRNA | AF088903.1 | - | 6, 7 | 829 | ||
AF160355.1 | - | 6, 7 | 817 | |||
AK077625.1 | - | 3, 4 | 536 | |||
AK080610.1 | - | 6, 7 | 820 | |||
AK166069.1 | - | 6, 7 | 822 | |||
AK167088.1 | - | 3, 4 | 545 | |||
AK172879.1 | - | 5, 6 | 734 | |||
BC014800.1 | - | 6, 7 | 802 | 4220980 |
Gene Symbol: Gm8615 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CB237974.1 | - | 1 | 50 |