Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gimap6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153175.3 | NP_694815.1 | 2 | 303 | ||
GenBank mRNA | AF503921.1 | - | 2 | 284 | ||
AK030902.1 | - | 2 | 273 | |||
AK050328.1 | - | 2 | 245 | |||
AK087275.1 | - | 2 | 273 | |||
AK160244.1 | - | 2 | 257 | |||
AK220304.1 | - | 2 | 237 | |||
BC028779.1 | - | 2 | 255 | 1196098 | ||
BU699766.1 | - | 2 | 303 |