Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C87436 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243741.1 | NP_001230670.1 | 7, 8 | 2099 | ||
NM_001243742.1 | NP_001230671.1 | 7, 8 | 1665 | |||
NM_146170.4 | NP_666282.3 | 7, 8 | 1606 | |||
GenBank mRNA | AK047127.1 | - | 7, 8 | 1607 | ||
AK077101.1 | - | 1486 | ||||
AK077503.1 | - | 7, 8 | 1986 | |||
AK170524.1 | - | 8, 9 | 1497 | |||
AK171511.1 | - | 8, 9 | 1497 | |||
BC025632.1 | - | 7, 8 | 1573 | 5321669 | ||
BC026926.1 | - | 8, 9 | 1730 | 3586097 | ||
BC031913.1 | - | 7, 8 | 1619 | 5376246 | ||
BU613064.1 | - | 3, 4 | 324 |