Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037727.2 | NP_001032816.1 | 4 | 749 | ||
NM_001286610.1 | NP_001273539.1 | 3 | 549 | |||
NM_175476.4 | NP_780685.2 | 4 | 420 | |||
GenBank mRNA | AK037710.1 | - | 4 | 482 | ||
AK048450.1 | - | 4 | 379 | |||
AK137444.1 | - | 4 | 408 | |||
AK137526.1 | - | 4 | 357 | |||
AK159598.1 | - | 3 | 550 | |||
AK159941.1 | - | 3 | 550 | |||
AK169897.1 | - | 4 | 726 | |||
AK172878.1 | - | 2 | 79 | |||
BC108400.1 | - | 1 | 30 | |||
BC138751.1 | - | 3 | 328 | 8861773 | ||
BC138752.1 | - | 3 | 328 | 8861774 | ||
CJ145934.1 | - | 4 | 421 |