Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172730.2 | NP_766318.2 | 8 | 866 | ||
GenBank mRNA | AK039818.1 | - | 8 | 845 | ||
AK046253.1 | - | 4 | 2712 | |||
AK081505.1 | - | 7 | 3511 | |||
AK156259.1 | - | 8 | 849 | |||
BC016607.1 | - | 6 | 634 | |||
BC051147.1 | - | 8 | 860 | 6595828 |