Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172731.2 | NP_766319.2 | 1 | 715 | ||
GenBank mRNA | AK220153.1 | - | 2 | 1189 | ||
BC060664.1 | - | 1 | 178 | |||
BC145502.1 | - | 2 | 529 | 9054065 | ||
BC150786.1 | - | 1 | 664 | 9087697 |