Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172731.2 | NP_766319.2 | 18 | 3997 | ||
GenBank mRNA | AK049341.1 | - | 5 | 671 | ||
AK086031.1 | - | 4 | 507 | |||
AK220153.1 | - | 19 | 4471 | |||
BC042732.1 | - | 14 | 1414 | |||
BC060664.1 | - | 18 | 3460 | |||
BC145502.1 | - | 19 | 3814 | 9054065 | ||
BC150786.1 | - | 18 | 3946 | 9087697 |