Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178703.4 | NP_848818.1 | 15 | 1949 | ||
GenBank mRNA | AK042956.1 | - | 15 | 1811 | ||
AK052971.1 | - | 15 | 1774 | |||
AK053883.1 | - | 15 | 1771 | |||
BC027111.1 | - | 2 | 144 | |||
BC059080.1 | - | 15 | 1800 | 6827121 | ||
M92378.1 | - | 14 | 1708 |