Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crebl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177687.3 | NP_808355.1 | 2 | 313 | ||
GenBank mRNA | AK045467.1 | - | 2 | 314 | ||
AK195782.1 | - | 2 | 87 | |||
BC109358.1 | - | 2 | 185 | 40057562 | ||
BC109359.1 | - | 2 | 201 | 40057564 |