Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk38l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172734.3 | NP_766322.1 | 7 | 784 | ||
GenBank mRNA | AK045971.1 | - | 5 | 521 | ||
AK052288.1 | - | 7 | 720 | |||
AY223819.1 | - | 7 | 706 | |||
BC062170.1 | - | 6 | 678 | 30135051 |