Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Caprin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301351.1 | NP_001288280.1 | 12 | 1389 | ||
NM_181541.4 | NP_853519.2 | 13 | 2046 | |||
GenBank mRNA | AK129460.1 | - | 12 | 1302 | ||
AK140744.1 | - | 13 | 2049 | |||
BC025636.1 | - | 13 | 2002 | |||
BC125373.1 | - | 13 | 1935 | 40129888 | ||
BC125375.1 | - | 13 | 1935 | 40129890 |