Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3hc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172735.2 | NP_766323.1 | 8 | 1049 | ||
GenBank mRNA | AJ537495.1 | - | 8 | 1041 | ||
AK088527.1 | - | 8 | 1050 | |||
AK146228.1 | - | 8 | 1064 | |||
AK151694.1 | - | 8 | 1045 | |||
AK167992.1 | - | 8 | 1064 | |||
AK168071.1 | - | 8 | 1059 | |||
BC024560.1 | - | 2 | 65 | |||
BC037445.1 | - | 8 | 1003 | |||
BC050141.1 | - | 8 | 1042 | 30011212 |