Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024698.2 | NP_001019869.1 | 1 | 67 | ||
GenBank mRNA | BC094929.1 | - | 1 | 97 | 6434004 |