Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Leng8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172736.3 | NP_766324.1 | 4 | 441 | ||
GenBank mRNA | AK034310.1 | - | 4 | 446 | ||
AK085304.1 | - | 4 | 439 | |||
AK170629.1 | - | 4 | 449 | |||
AK200008.1 | - | 1 | 78 | |||
BC041775.1 | - | 4 | 413 | |||
BC042658.1 | - | 4 | 429 | 5365041 | ||
BC066768.1 | - | 4 | 432 | 6407146 | ||
CF536188.1 | - | 4 | 441 |