Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172739.4 | NP_766327.3 | 2 | 2500 | ||
GenBank mRNA | AK147326.1 | - | 2 | 2500 | ||
AK147688.1 | - | 2 | 2500 | |||
AK161796.1 | - | 2 | 2095 | |||
AK173242.1 | - | 1 | 1301 | |||
BC139459.1 | - | 3 | 2278 | 8862481 | ||
BC139461.1 | - | 3 | 2137 | 8862483 |