Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mark4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172279.1 | NP_758483.1 | 3 | 271 | ||
GenBank mRNA | AK146784.1 | - | 4 | 606 | ||
AY151083.1 | - | 3 | 271 |