Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpul1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144922.2 | NP_659171.1 | 4 | 708 | ||
NM_178089.2 | NP_835190.1 | 4 | 689 | |||
GenBank mRNA | AK042090.1 | - | 4 | 689 | ||
AK087545.1 | - | 4 | 678 | |||
AK143666.1 | - | 4 | 751 | |||
AK155604.1 | - | 4 | 719 | |||
AK203240.1 | - | 1 | 44 | |||
BC021506.1 | - | 4 | 732 | 5347099 | ||
BC078642.1 | - | 4 | 632 | 6329630 |