Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163702.1 | NP_001157174.1 | 4 | 757 | ||
NM_207238.3 | NP_997121.1 | 5 | 752 | |||
GenBank mRNA | AK053292.1 | - | 5 | 754 | ||
AK140275.1 | - | 4 | 755 | |||
AK158463.1 | - | 4 | 758 | |||
AK169614.1 | - | 4 | 842 | |||
BC075656.1 | - | 4 | 720 | 30540609 |