Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alkbh6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198027.2 | NP_932144.2 | 7 | 663 | ||
GenBank mRNA | BC029805.1 | - | 7 | 615 | 4952089 | |
BC080310.1 | - | 5 | 786 |