Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syne4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153577.2 | NP_705805.1 | 5 | 923 | ||
GenBank mRNA | AA407649.1 | - | 3 | 394 | ||
BC004761.1 | - | 5 | 872 | 3155758 | ||
BC023803.1 | - | 5 | 910 | 5352953 | ||
BC056649.1 | - | 5 | 862 | 5036575 |