Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrfn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175478.2 | NP_780687.1 | 2 | 1123 | ||
GenBank mRNA | AK040916.1 | - | 2 | 1124 | ||
BC066999.1 | - | 3 | 1046 | 6418622 | ||
BC094593.1 | - | 2 | 1085 | 5708704 | ||
DQ078787.1 | - | 1 | 267 |