Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289670.1 | NP_001276599.1 | 18 | 1909 | ||
NM_001289682.1 | NP_001276611.1 | 19 | 1992 | |||
NM_178252.3 | NP_839983.1 | 19 | 2019 | |||
GenBank mRNA | AK142180.1 | - | 18 | 1910 | ||
AY217764.1 | - | 18 | 1852 | |||
BC061471.1 | - | 12 | 1196 | 5698780 | ||
BC065086.1 | - | 19 | 1968 | 6850475 | ||
BC065166.1 | - | 19 | 1966 | 6836510 | ||
BC066047.1 | - | 19 | 1944 | 6822191 |