Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4931406P16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172741.2 | NP_766329.1 | 6 | 1518 | ||
GenBank mRNA | AK076994.1 | - | 6 | 1519 | ||
AK129125.1 | - | 5 | 496 | |||
AK170198.1 | - | 7 | 734 | |||
BC060233.1 | - | 6 | 1319 | 6852905 |