Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Siglecf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271019.1 | NP_001257948.1 | 8 | 1474 | ||
NM_145581.2 | NP_663556.1 | 9 | 1576 | |||
GenBank mRNA | AF293371.1 | - | 9 | 1512 | ||
AK087658.1 | - | 8 | 1639 | |||
AK146203.1 | - | 8 | 1613 | |||
AK155469.1 | - | 9 | 1525 | |||
AY210401.1 | - | 8 | 1472 | |||
BC145038.1 | - | 7 | 1405 | 9053586 | ||
BC145987.1 | - | 8 | 1507 | 40131290 |