Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Luzp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178705.5 | NP_848820.4 | 12 | 1667 | ||
GenBank mRNA | AK032204.1 | - | 12 | 1324 | ||
AK034077.1 | - | 12 | 1285 | |||
AK034240.1 | - | 12 | 1319 | |||
AY164459.1 | - | 12 | 1664 | |||
BC075646.1 | - | 12 | 1297 | 30620475 |