Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Siglech | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178706.4 | NP_848821.2 | 1 | 201 | ||
GenBank mRNA | AK032377.1 | - | 1 | 154 | ||
AK047813.1 | - | 1 | 119 | |||
AK080658.1 | - | 1 | 195 | |||
BC098229.1 | - | 1 | 140 | 1430130 | ||
BC120501.1 | - | 1 | 143 | 40129424 | ||
BC144741.1 | - | 1 | 143 | 9053289 | ||
BC144742.1 | - | 1 | 143 | 9053290 | ||
BC144743.1 | - | 1 | 143 | 9053291 | ||
DQ157764.1 | - | 1 | 120 | |||
DQ208935.1 | - | 1 | 146 |