Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtmr10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172742.2 | NP_766330.2 | 13 | 1435 | ||
GenBank mRNA | AK028584.1 | - | 1 | 60 | ||
BC041672.1 | - | 13 | 1407 | |||
BC055074.1 | - | 11 | 1099 | 30052208 | ||
BC058196.1 | - | 6 | 537 | |||
CB600359.1 | - | 1 | 106 |