Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002686.4 | NP_002677.1 | 2 | 318 | ||
NR_073461.1 | 2 | 183 | ||||
XM_011524909.2 | XP_011523211.1 | 2 | 426 | |||
GenBank mRNA | BC037246.1 | - | 2 | 517 | 5175571 | |
CR541815.1 | - | 2 | 295 | |||
J03727.1 | - | 2 | 301 |