Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010826.3 | NP_001010826.2 | 3 | 415 | ||
NM_001012434.3 | NP_001012436.1 | 3 | 448 | |||
NM_001136235.1 | NP_001129707.1 | 3 | 432 | |||
GenBank mRNA | AK145279.1 | - | 3 | 416 | ||
AY899809.1 | - | 3 | 448 | |||
BC024588.1 | - | 3 | 287 | 5066294 | ||
BG088171.2 | - | 3 | 432 | |||
BY735425.1 | - | 3 | 416 | |||
CK791017.1 | - | 3 | 336 |