Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002687.3 | NP_002678.2 | 9 | 2070 | ||
GenBank mRNA | AK223612.1 | - | 9 | 2005 | ||
AK292579.1 | - | 9 | 2054 | |||
AK300164.1 | - | 2 | 1319 | |||
AK303136.1 | - | 5 | 1661 | |||
AK304622.1 | - | 1 | 1175 | |||
BC062602.1 | - | 9 | 1981 | 5491466 | ||
BM850066.1 | - | 1 | 547 | |||
U59479.1 | - | 1 | 121 | |||
U77718.1 | - | 9 | 2081 | |||
Y09703.1 | - | 9 | 1986 |