Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2210018M11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172280.2 | NP_758484.2 | 11 | 1662 | ||
GenBank mRNA | AK032985.1 | - | 10 | 1647 | ||
BC039956.1 | - | 11 | 1653 | |||
BC066050.1 | - | 9 | 1522 | |||
BC089304.1 | - | 11 | 1662 | 6854147 |