Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002687.3 | NP_002678.2 | 9 | 917 | ||
GenBank mRNA | AK223612.1 | - | 9 | 852 | ||
AK292579.1 | - | 9 | 901 | |||
AK300164.1 | - | 2 | 166 | |||
AK303136.1 | - | 5 | 508 | |||
AK304622.1 | - | 1 | 22 | |||
BC062602.1 | - | 9 | 828 | 5491466 | ||
U77718.1 | - | 9 | 847 | |||
Y09703.1 | - | 9 | 827 |