Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291358.1 | NP_001278287.1 | 7 | 1032 | ||
NM_145583.3 | NP_663558.1 | 7 | 898 | |||
GenBank mRNA | AK134287.1 | - | 7 | 915 | ||
AK146442.1 | - | 7 | 856 | |||
AK152420.1 | - | 6 | 851 | |||
AK152931.1 | - | 6 | 570 | |||
AK159933.1 | - | 7 | 873 | |||
AK159950.1 | - | 7 | 871 | |||
AK162960.1 | - | 8 | 736 | |||
BC004794.1 | - | 7 | 852 | 3588752 |