Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007106.3 | NP_009037.1 | 2 | 1225 | ||
GenBank mRNA | AF044221.1 | - | 2 | 184 | ||
AK056580.1 | - | 2 | 1120 | |||
AK307361.1 | - | 2 | 169 | |||
AK309253.1 | - | 2 | 154 | |||
AK311846.1 | - | 2 | 268 | |||
AL080177.1 | - | 2 | 189 | |||
BC059385.1 | - | 2 | 565 | 5287541 | ||
CR533458.1 | - | 2 | 80 |