Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ipo7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181517.3 | NP_852658.2 | 19, 20 | 2180 | ||
GenBank mRNA | AK039534.1 | - | 19, 20 | 2332 | ||
AY316153.1 | - | 19, 20 | 2180 | |||
BC053524.1 | - | 16, 17 | 1863 | |||
BC064825.1 | - | 10, 11 | 1136 |