Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spon1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145584.2 | NP_663559.1 | 7, 8 | 1218 | ||
GenBank mRNA | AK146506.1 | - | 7, 8 | 1043 | ||
AK173021.1 | - | 7, 8 | 885 | |||
BC020531.1 | - | 7, 8 | 1088 | 4221758 |