Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007106.3 | NP_009037.1 | 3 | 1327 | ||
GenBank mRNA | AF044221.1 | - | 3 | 286 | ||
AK056580.1 | - | 3 | 1222 | |||
AK307361.1 | - | 3 | 271 | |||
AK309253.1 | - | 3 | 256 | |||
AK311846.1 | - | 3 | 370 | |||
AL080177.1 | - | 3 | 291 | |||
BC044582.1 | - | 3 | 770 | 5262176 | ||
BC059385.1 | - | 3 | 667 | 5287541 | ||
CR533458.1 | - | 3 | 182 |